Novus Biologicals
Manufacturer Code:NBP191654
Catalog # NBP191654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:YHMYRERGGELLVHTGFLGSSQDRSAYQTIDSAEAPADPFAVPEGRSQDARGY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: angiotensin II receptor-associated protein; angiotensin II receptor-associated protein AT1 receptor-associated protein ATI receptor-associated protein MGC29646 type I receptor-associated protein type-1 angiotensin II receptor-associated protein; angiotensin II, type I receptor-associated protein; AT1 receptor-associated protein; ATI receptor-associated protein; Type-1 angiotensin II receptor-associated protein
Gene Aliases: AGTRAP; ATRAP
UniProt ID: (Human) Q6RW13
Entrez Gene ID: (Human) 57085
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.