Invitrogen
Manufacturer Code:PA520813
Catalog # PIPA520813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide from near the center of human AGTR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: angiotensin II receptor, type 2; angiotensin II type 2 receptor; Angiotensin II type-2 receptor; angiotensin receptor 2; AT2; AT2 ATGR2 MRX88 angiotensin II type-2 receptor type-2 angiotensin II receptor; AT2 receptor; Type-2 angiotensin II receptor
Gene Aliases: AGTR2; AI316812; AT2; AT2-R; AT2R; AT<sub>2</sub>R; ATGR2; AW107640; MRX88
UniProt ID: (Human) P50052, (Rat) P35351, (Mouse) P35374
Entrez Gene ID: (Human) 186, (Rat) 24182, (Mouse) 11609
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.