Invitrogen
Manufacturer Code:MA537535
Catalog # PIMA537535
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 338CT11.2.5 |
Immunogen | KLH conjugated synthetic peptide between 211-240 amino acids from the Central region of human AGTR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 1810074K20Rik AG2S Agtr1 Agtr1a Agtr-1a AGTR1B AI551199 angiotensin II receptor type 1 angiotensin II receptor type 1 angiotensin II receptor type 1a Angiotensin II type-1 receptor Angiotensin II type-1A receptor angiotensin receptor 1 angiotensin receptor 1a Angtr-1a AT1 At1a AT1AR AT1B AT1BR AT1R AT2R1 AT2R1A AT2R1B HAT1R Type-1 angiotensin II receptor type-1A angiotensin II receptor type-1B angiotensin II receptor vascular type-1 angiotensin II receptor; angiotensin II receptor, type 1; Angiotensin II type-1 receptor; AT1; AT1AR; AT1BR; Type-1 angiotensin II receptor; type-1B angiotensin II receptor
Gene Aliases: AG2S; AGTR1; AGTR1A; AGTR1B; AT1; AT1AR; AT1B; AT1BR; AT1R; AT2R1; AT2R1B; HAT1R
UniProt ID: (Human) P30556
Entrez Gene ID: (Human) 185
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.