Invitrogen
Manufacturer Code:PA518587
Catalog # PIPA518587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (EIQKNKPRNDDIFK) corresponding to the internal amino acids of AGTR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AGTR1 AT1 angiotensin II receptor type 1 AG2S AGTR1A AGTR1B AT1B AT2R1 AT2R1A AT2R1B HAT1R angiotensin receptor 1 angiotensin receptor 1B type-1B angiotensin II receptor; angiotensin II receptor, type 1; Angiotensin II type-1 receptor; Angiotensin II type-1A receptor; angiotensin receptor 1; angiotensin receptor 1a; AT1; AT1A; AT1AR; AT1BR; Type-1 angiotensin II receptor; Type-1A angiotensin II receptor; type-1B angiotensin II receptor; vascular type-1 angiotensin II receptor
Gene Aliases: 1810074K20Rik; AG2S; Agtr-1a; AGTR1; AGTR1A; AGTR1B; AI551199; Angtr-1a; AT1; AT1A; AT1AR; AT1B; AT1BR; AT1R; AT2R1; AT2R1A; AT2R1B; HAT1R
UniProt ID: (Human) P30556, (Rat) P25095, (Mouse) P29754
Entrez Gene ID: (Human) 185, (Rat) 24180, (Mouse) 11607
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.