Abnova
Manufacturer Code:PAB24974
Catalog # 89-126-650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 150-200 of human AGTR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of AGTR1.
For Research Use Only
Protein Aliases: AG2S; AGTR1A; AGTR1B; angiotensin II receptor, type 1; Angiotensin II type-1 receptor; AT1; AT1AR; AT1B; AT1BR; AT1R; AT2R1; AT2R1A; AT2R1B; HAT1R; Type-1 angiotensin II receptor; type-1B angiotensin II receptor
Gene Aliases: AG2S; AGTR1; AGTR1A; AGTR1B; AT1; AT1AR; AT1B; AT1BR; AT1R; AT2R1; AT2R1B; HAT1R
UniProt ID: (Human) P30556
Entrez Gene ID: (Human) 185
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.