Abnova
Manufacturer Code:H00000185K
Catalog # 89-200-029
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human AGTR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human AGTR1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: AG2S AGTR1A AGTR1B AT1 AT1B AT1R AT2R1 AT2R1A AT2R1B HAT1R; angiotensin II receptor, type 1; Angiotensin II type-1 receptor; AT1; AT1AR; AT1BR; Type-1 angiotensin II receptor; type-1B angiotensin II receptor
Gene Aliases: AG2S; AGTR1; AGTR1A; AGTR1B; AT1; AT1AR; AT1B; AT1BR; AT1R; AT2R1; AT2R1B; HAT1R
UniProt ID: (Human) P30556
Entrez Gene ID: (Human) 185
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.