R&D Systems
Manufacturer Code:FAB3659N100UG
Catalog # FAB3659N100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 364805 |
Immunogen | HEK293 human embryonic kidney cell line transfected with human AGTR-2 Met1-Ser363 Accession # P50052 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: AGTR2 Angiostensin Receptor angiotensin II receptor type 2 Angiotensin II type-2 receptor angiotensin receptor 2 AT2 AT2ATGR2 AT2R MRX88 MRX88type-2 angiotensin II receptor; angiotensin II receptor, type 2; Angiotensin II type-2 receptor; AT2; Type-2 angiotensin II receptor
Gene Aliases: AGTR2; AT2; ATGR2; MRX88
UniProt ID: (Human) P50052
Entrez Gene ID: (Human) 186
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.