R&D Systems
Manufacturer Code:MAB102441100
Catalog # MAB10244110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1010106 |
Immunogen | Synthetic peptide containing human AGTR-1 Accession # P30556 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: AG2S AGTR1 AGTR1AAngiotensin II type-1 receptor Angiostensin Receptor angiotensin II receptor type 1 angiotensin receptor 1 angiotensin receptor 1B AT1 AT1AR AT1AT1AR AT1B AT1BR AT1R AT2R1 AT2R1A AT2R1AGTR1B AT2R1BAT1BR HAT1R type-1 angiotensin II receptor type-1B angiotensin II receptor; angiotensin II receptor, type 1; Angiotensin II type-1 receptor; AT1; AT1AR; AT1BR; Type-1 angiotensin II receptor; type-1B angiotensin II receptor
Gene Aliases: AG2S; AGTR1; AGTR1A; AGTR1B; AT1; AT1AR; AT1B; AT1BR; AT1R; AT2R1; AT2R1B; HAT1R
UniProt ID: (Human) P30556
Entrez Gene ID: (Human) 185
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.