OriGene
Manufacturer Code:TA800287
Catalog # 50-167-8229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9C8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 368-753 of human AGTPBP1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP/GTP-binding protein 1; carboxypeptidase-tubulin; CCP1 NNA1; Cytosolic carboxypeptidase 1; Nervous system nuclear protein induced by axotomy protein 1 homolog; soluble carboxypeptidase; tubulinyl-Tyr carboxypeptidase; tyrosine carboxypeptidase
Gene Aliases: AGTPBP1; CCP1; KIAA1035; NNA1
UniProt ID: (Human) Q9UPW5
Entrez Gene ID: (Human) 23287
Molecular Function: G-protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.