Novus Biologicals
Manufacturer Code:NBP246502
Catalog # NBP246502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9A3 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 368-753 of human AGTPBP1 (NP_056054) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP/GTP binding protein 1 ATP/GTP-binding protein 1 CCP1 DKFZp686M20191 EC 3.4.17 EC 3.4.17.- KIAA1035NNA1cytosolic carboxypeptidase 1 Nervous system nuclear protein induced by axotomy protein 1 homolog Nna1; ATP/GTP-binding protein 1; carboxypeptidase-tubulin; Cytosolic carboxypeptidase 1; Nervous system nuclear protein induced by axotomy protein 1 homolog; soluble carboxypeptidase; tubulinyl-Tyr carboxypeptidase; tyrosine carboxypeptidase
Gene Aliases: AGTPBP1; CCP1; KIAA1035; NNA1
UniProt ID: (Human) Q9UPW5
Entrez Gene ID: (Human) 23287
Molecular Function:
G-protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.