Invitrogen
Manufacturer Code:PA544291
Catalog # PIPA544291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human AGTPBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP/GTP-binding protein 1; carboxypeptidase-tubulin; CCP1; Cytosolic carboxypeptidase 1; Cytosolic carboxypeptidase 1-like protein; Nervous system nuclear protein induced by axotomy protein 1; Nervous system nuclear protein induced by axotomy protein 1 homolog; nuclear ATP/GTP-binding protein; Purkinje cell degeneration; soluble carboxypeptidase; tubulinyl-Tyr carboxypeptidase; tyrosine carboxypeptidase
Gene Aliases: 1700020N17Rik; 2310001G17Rik; 2900054O13Rik; 4930445M19Rik; 5730402G09Rik; AGTPBP1; CCP1; KIAA1035; nmf243; NNA1; pcd; si:ch73-236b16.2
UniProt ID: (Human) Q9UPW5, (Mouse) Q641K1, (Zebrafish) Q4U2V3
Entrez Gene ID: (Pig) 100518857, (Dog) 484146, (Human) 23287, (Rabbit) 103352412, (Horse) 100061665, (Guinea pig) 100726862, (Mouse) 67269, (Rat) 290986, (Zebrafish) 554164
Molecular Function:
G-protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.