Abnova
Manufacturer Code:PAB5053
Catalog # 89-131-706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to human AGT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of AGT.
For Research Use Only
Protein Aliases: alpha-1 antiproteinase, antitrypsin; Ang I; Ang II; Ang III; Ang IV; Angiotensin 1-10; Angiotensin 1-4; Angiotensin 1-5; Angiotensin 1-7; Angiotensin 1-8; Angiotensin 1-9; Angiotensin 2-8; Angiotensin 3-8; Angiotensin I; Angiotensin II; Angiotensin III; Angiotensin IV; Angiotensin-1; Angiotensin-2; Angiotensin-3; Angiotensin-4; Angiotensinogen; angiotensinogen (serpin peptidase inhibitor, clade A, member 8); ANHU; Des-Asp[1]-angiotensin II; FLJ92595; FLJ97926; pre-angiotensinogen; serine (or cysteine) proteinase inhibitor; Serpin A8; serpin peptidase inhibitor, clade A, member 8; SERPINA8
Gene Aliases: AGT; ANHU; SERPINA8
UniProt ID: (Human) P01019
Entrez Gene ID: (Human) 183
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.