Invitrogen
Manufacturer Code:PA576591
Catalog # PIPA576591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human AGPS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADAS ADPS RCDP3 ADAP-S ADHAPS ALDHPSY; Aging-associated gene 5 protein; aging-associated protein 5; Alkyl-DHAP synthase; Alkyldihydroxyacetonephosphate synthase, peroxisomal; Alkylglycerone-phosphate synthase
Gene Aliases: 5832437L22; 9930035G10Rik; AAG5; ADAP-S; Adaps; ADAS; ADHAPS; ADPS; AGPS; ALDHPSY; AW123847; bs2; RCDP3
UniProt ID: (Human) O00116, (Mouse) Q8C0I1
Entrez Gene ID: (Human) 8540, (Mouse) 228061
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.