Abnova
Manufacturer Code:PAB18829
Catalog # 89-120-388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids 462-632 of human AGPS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant AGPS.
For Research Use Only
Protein Aliases: ADAP-S; ADAS; ADHAPS; ADPS; Aging-associated gene 5 protein; aging-associated protein 5; ALDHPSY; Alkyl-DHAP synthase; Alkyldihydroxyacetonephosphate synthase, peroxisomal; Alkylglycerone-phosphate synthase; DKFZp762O2215; FLJ99755
Gene Aliases: AAG5; ADAP-S; ADAS; ADHAPS; ADPS; AGPS; ALDHPSY; RCDP3
UniProt ID: (Human) O00116
Entrez Gene ID: (Human) 8540
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.