Proteintech
Manufacturer Code:206031AP150UL
Catalog # 50-173-2984
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AGPAT9 Fusion Protein Ag14579 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1 AGP acyltransferase 9 1 AGPAT 9 AGPAT8 AGPAT9 GPAT 3 GPAT3 hGPAT3 HMFN0839 LPAAT theta MAG 1 MAG1 UNQ2753/PRO6492; 1-acyl-sn-glycerol-3-phosphate O-acyltransferase 10; 1-acyl-sn-glycerol-3-phosphate O-acyltransferase 9; 1-acylglycerol-3-phosphate O-acyltransferase 8; 1-acylglycerol-3-phosphate O-acyltransferase 9; 1-AGP acyltransferase 9; 1-AGPAT 9; Acyl-CoA:glycerol-3-phosphate acyltransferase 3; AGPAT 10; endoplasmic reticulum associated GPAT; Glycerol-3-phosphate acyltransferase 3; GPAT-3; GPAT3; hGPAT3; LPAAT-theta; Lung cancer metastasis-associated protein 1; Lysophosphatidic acid acyltransferase theta; lysophosphatidic acid acyltransferase, theta; MAG-1; mGPAT3; testis secretory sperm-binding protein Li 213e
Gene Aliases: 1-AGPAT; 1-AGPAT 9; 4933407I02Rik; 4933408F15; 9; A230097K15Rik; AGPAT 10; AGPAT10; AGPAT8; AGPAT9; GPAT-3; GPAT3; HMFN0839; LPAAT-theta; MAG1; mGPAT3; UNQ2753/PRO6492
UniProt ID: (Rat) Q4V8J4, (Mouse) Q8C0N2, (Human) Q53EU6
Entrez Gene ID: (Rat) 305166, (Mouse) 231510, (Human) 84803
Molecular Function: acyltransferase annexin calcium-binding protein calmodulin intracellular calcium-sensing protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.