Proteintech
Manufacturer Code:167621AP150UL
Catalog # 50-173-1162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AGPAT6 Fusion Protein Ag10294 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1 AGP acyltransferase 6 1 AGPAT 6 AGPAT6 GPAT4 LPAAT zeta LPAATZ TSARG7; 1-acyl-sn-glycerol-3-phosphate acyltransferase zeta; 1-acylglycerol-3-phosphate O-acyltransferase 6; 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta); 1-AGP acyltransferase 6; 1-AGPAT 6; Acyl-CoA:glycerol-3-phosphate acyltransferase 4; Glycerol-3-phosphate acyltransferase 4; glycerol-3-phosphate acyltransferase 6; glycerol-3-phosphate acyltransferase-4; GPAT4; LPAAT-zeta; Lysophosphatidic acid acyltransferase zeta; putative lysophosphatidic acid acyltransferase; Testis spermatogenesis apoptosis-related protein 7; TSARG7
Gene Aliases: 1-AGPAT 6; AGPAT6; AU041707; AW545732; GPAT4; LPAAT-zeta; LPAATZ; RGD1310520; TSARG7; UNQ551/PRO1108
UniProt ID: (Mouse) Q8K2C8, (Human) Q86UL3
Entrez Gene ID: (Rat) 290843, (Mouse) 102247, (Human) 137964
Molecular Function:
acyltransferase
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.