Invitrogen
Manufacturer Code:PA5103511
Catalog # PIPA5103511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AGPAT1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-acyl-sn-glycerol-3-phosphate acyltransferase alpha; 1-acylglycerol-3-phosphate O-acyltransferase 1; 1-acylglycerol-3-phosphate O-acyltransferase 1 (acetoacetly Coenzyme A thiolase); 1-acylglycerol-3-phosphate O-acyltransferase 1 (lysophosphatidic acid acyltransferase, alpha); 1-acylglycerol-3-phosphate O-acyltransferase 1 1-acylglycerol-3-phosphate O-acyltransferase 1 (acetoacetly Coenzyme A thiolase) 1-acylglycerol-3-phosphate O-acyltransferase 1 (lysophosphatidic acid acyltransferase alpha) 1-acyl-sn-glycerol-3-phosphate acyltransferase alpha 1-AGP 1-AGP acyltransferase 1 1-AGPAT 1-AGPAT 1 1-AGPAT1 Agpat1 AW047140 G15 LPAATA LPAAT-alpha Lysophosphatidic acid acyltransferase alpha lysophospholipid acyltransferase Protein G15; 1-AGP acyltransferase 1; 1-AGPAT 1; LPAAT-alpha; Lysophosphatidic acid acyltransferase alpha; lysophospholipid acyltransferase; Protein G15
Gene Aliases: 1-AGP; 1-AGPAT; 1-AGPAT1; AGPAT1; AW047140; G15; LPAAT-alpha; LPAATA
UniProt ID: (Human) Q99943, (Mouse) O35083
Entrez Gene ID: (Human) 10554, (Rat) 406165, (Mouse) 55979
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.