Invitrogen
Manufacturer Code:MA537960
Catalog # PIMA537960
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0189 |
Immunogen | A synthesized peptide derived from human Argonaute-2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 1110029L17Rik 2310051F07Rik AGO1 Ago2 AI225898 AL022874 argonaute 2 argonaute 2 RISC catalytic component argonaute RISC catalytic component 2 argonaute RISC catalytic subunit 2 Argonaute1 argonaute2 argonaute2 {ECO:0000255 AW546247 CTA-204B4.6 eIF2C eIF2C 1 eIF-2C 1 eIF2C 2 eIF-2C 2 eIF2C 2 {ECO:0000255 eIF-2C 2 {ECO:0000255 EIF2C1 Eif2c2 ENSMUSG00000072493 Eukaryotic translation initiation factor 2C 2 eukaryotic translation initiation factor 2C 2 {ECO:0000255 eukaryotic translation initiation factor 2C 2 GERp95 Gm10365 golgi ER protein 95 kDa hAgo1 hAgo2 HAMAP-Rule:MF_03031} Kiaa4215 mAgo2 mKIAA4215 PAZ Piwi domain protein Piwi/Argonaute family protein meIF2C2 PPD Protein argonaute-1 protein argonaute-2 Protein slicer Q10; argonaute 2; Argonaute RISC catalytic component 2; Argonaute2; argonaute2 {ECO:0000255|HAMAP-Rule:MF_03031}; CTA-204B4.6; eIF-2C 2; eIF-2C 2 {ECO:0000255|HAMAP-Rule:MF_03031}; eIF2C 2; eIF2C 2 {ECO:0000255|HAMAP-Rule:MF_03031}; Eukaryotic translation initiation factor 2C 2; eukaryotic translation initiation factor 2C 2 {ECO:0000255|HAMAP-Rule:MF_03031}; eukaryotic translation initiation factor 2C, 2; GERp95; Golgi ER protein 95 kDa; hAgo2; mAgo2; PAZ Piwi domain protein; Piwi/Argonaute family protein meIF2C2; PPD; Protein argonaute-2; Protein slicer
Gene Aliases: 1110029L17Rik; 2310051F07Rik; AGO2; AI225898; AL022874; AW546247; EIF2C2; ENSMUSG00000072493; Gerp95; Gm10365; Kiaa4215; mKIAA4215; Q10
UniProt ID: (Human) Q9UKV8, (Rat) Q9QZ81, (Mouse) Q8CJG0
Entrez Gene ID: (Human) 27161, (Rat) 59117, (Mouse) 239528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.