Invitrogen
Manufacturer Code:MA529649
Catalog # PIMA529649
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 36 |
Immunogen | Recombit Mouse AGO2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: argonaute 2; Argonaute RISC catalytic component 2; Argonaute2; eIF-2C 2; Eukaryotic translation initiation factor 2C 2; eukaryotic translation initiation factor 2C, 2; mAgo2; PAZ Piwi domain protein PPD argonaute 2 argonaute2 eIF-2C 2 eIF2C 2 hAgo2 protein argonaute-2 protein slicer; Piwi/Argonaute family protein meIF2C2; Protein argonaute-2; Protein slicer
Gene Aliases: 1110029L17Rik; 2310051F07Rik; Ago2; AI225898; AL022874; AW546247; Eif2c2; ENSMUSG00000072493; Gerp95; Gm10365; Kiaa4215; mKIAA4215
UniProt ID: (Mouse) Q8CJG0
Entrez Gene ID: (Mouse) 239528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.