Invitrogen
Manufacturer Code:PA572107
Catalog # PIPA572107
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 27-59 amino acids from the N-terminal region of human EIF2C2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: argonaute 2; Argonaute RISC catalytic component 2; Argonaute2; CTA-204B4.6; eIF-2C 2; eIF2C 2; Eukaryotic translation initiation factor 2C 2; eukaryotic translation initiation factor 2C, 2; hAgo2; HAMAP-Rule:MF&lowbar03031} Ago2 Eif2c2 Kiaa4215; HAMAP-Rule:MF&lowbar03031} Argonaute RISC catalytic component 2 Eukaryotic translation initiation factor 2C 2 {ECO:0000255; HAMAP-Rule:MF&lowbar03031} Argonaute2 {ECO:0000255; HAMAP-Rule:MF&lowbar03031} eIF-2C 2 {ECO:0000255; HAMAP-Rule:MF&lowbar03031} eIF2C 2 {ECO:0000255; HAMAP-Rule:MF&lowbar03031} mAgo2 3126n2 {ECO:0000255; HAMAP-Rule:MF&lowbar03031} Piwi/argonaute family protein meIF2C2 Protein slicer {ECO:0000255; PAZ Piwi domain protein; PPD; Protein argonaute-2; Protein argonaute-2 {ECO:0000255; Protein slicer
Gene Aliases: AGO2; EIF2C2; Q10
UniProt ID: (Human) Q9UKV8
Entrez Gene ID: (Human) 27161
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.