Invitrogen
Manufacturer Code:PA566129
Catalog # PIPA566129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human AGO2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: argonaute 2; Argonaute RISC catalytic component 2; Argonaute2; CTA-204B4.6; eIF-2C 2; eIF2C 2; Eukaryotic translation initiation factor 2C 2; eukaryotic translation initiation factor 2C, 2; hAgo2; PAZ Piwi domain protein; PPD; Protein argonaute-2; Protein argonaute-2 Argonaute2 hAgo2 Argonaute RISC catalytic component 2 Eukaryotic translation initiation factor 2C eIF-2C 2 eIF2C PAZ Piwi domain protein PPD AGO2 EIF2C2 Protein argonaute-1 Argonaute1 hAgo1 Argonaute RISC catalytic component 1 Eukaryotic translation initiation factor 2C 1 eIF-2C 1 eIF2C 1 Putative RNA-binding protein Q99 AGO1 EIF2C1; Protein slicer
Gene Aliases: AGO2; EIF2C2; Q10
UniProt ID: (Human) Q9UKV8
Entrez Gene ID: (Human) 27161
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.