Novus Biologicals
Manufacturer Code:NBP17040420UL
Catalog # NBP17040420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to TMEM195 (transmembrane protein 195) The peptide sequence was selected from the middle region of TMEM195)(50ug). Peptide sequence AGHQTHHSSEDYNLSTALRQSVLQIYTSWIFYSPLALFIPPSVYAVHLQF. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alkylglycerol monooxygenase; alkylglycerol monooxygenase transmembrane protein 195; glyceryl-ether monooxygenase; Transmembrane protein 195
Gene Aliases: AGMO; TMEM195
UniProt ID: (Human) Q6ZNB7
Entrez Gene ID: (Human) 392636
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.