Invitrogen
Manufacturer Code:PA528566
Catalog # PIPA528566
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 235 of Human Acylglycerol kinase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylglycerol kinase, mitochondrial; hAGK; HsMuLK; MTDPS10 MULK acylglycerol kinase mitochondrial hAGK hsMuLK multi-substrate lipid kinase multiple substrate lipid kinase; MuLK; multi-substrate lipid kinase; Multiple substrate lipid kinase
Gene Aliases: 2610037M15Rik; 6720408I04Rik; AGK; AI465370; CATC5; CTRCT38; MTDPS10; MULK
UniProt ID: (Human) Q53H12, (Mouse) Q9ESW4
Entrez Gene ID: (Human) 55750, (Mouse) 69923
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.