Invitrogen
Manufacturer Code:PA552599
Catalog # PIPA552599
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human AGFG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arf-GAP domain and FG repeat-containing protein 1; arf-GAP domain and FG repeats-containing protein 1; HIV-1 Rev binding protein; HIV-1 Rev-binding protein; HIV-1 Rev-binding protein homolog; HRB RAB RIP HIV-1 Rev binding protein HIV-1 Rev-binding protein Rab Rev/Rex activation domain-binding protein arf-GAP domain and FG repeats-containing protein 1 hRIP Rev interacting protein nucleoporin-like protein RIP rev-interacting protein rev/Rex activation dom; hRIP, Rev interacting protein; Nucleoporin-like protein RIP; Rab, Rev/Rex activation domain-binding protein; Rev-interacting protein; Rev/Rex activation domain-binding protein
Gene Aliases: AGFG1; Agfg1-ps1; AU045498; C130049H11Rik; C85612; D730048C23Rik; HRB; RAB; RIP
UniProt ID: (Human) P52594, (Mouse) Q8K2K6, (Rat) Q4KLH5
Entrez Gene ID: (Human) 3267, (Mouse) 15463, (Rat) 363266
Molecular Function:
G-protein modulator
enzyme modulator
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.