Invitrogen
Manufacturer Code:PA530234
Catalog # PIPA530234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 18 and 305 of Human RIP/Rab |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arf-GAP domain and FG repeat-containing protein 1; arf-GAP domain and FG repeats-containing protein 1; HIV-1 Rev binding protein; HIV-1 Rev-binding protein; HRB RAB RIP HIV-1 Rev binding protein HIV-1 Rev-binding protein Rab Rev/Rex activation domain-binding protein arf-GAP domain and FG repeats-containing protein 1 hRIP Rev interacting protein nucleoporin-like protein RIP rev-interacting protein rev/Rex activation dom; hRIP, Rev interacting protein; NEUG(55-78); Neurogranin; neurogranin (protein kinase C substrate, RC3); Ng; Nucleoporin-like protein RIP; postsynaptic spine 1; protein kinase C substrate; Rab, Rev/Rex activation domain-binding protein; RC3; Rev-interacting protein; Rev/Rex activation domain-binding protein
Gene Aliases: 0710001B06Rik; AGFG1; AI838505; HRB; NG; NG/RC3; Nrgn; Pss1; R75334; RAB; RC3; RIP
UniProt ID: (Human) P52594, (Mouse) P60761
Entrez Gene ID: (Human) 3267, (Mouse) 64011
Molecular Function: G-protein modulator enzyme modulator nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.