Abnova
Manufacturer Code:MAB1724
Catalog # 89-110-595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C5 |
Immunogen | Recombinant protein corresponding to human AGER. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant AGER.
For Research Use Only
Protein Aliases: Advanced glycosylation end product-specific receptor; MGC22357; RAGE; RAGE isoform NtRAGE-delta; RAGE isoform sRAGE-delta; receptor for advanced glycation end-products variant 20; Receptor for advanced glycosylation end products
Gene Aliases: AGER; RAGE; SCARJ1
UniProt ID: (Human) Q9H2X7
Entrez Gene ID: (Human) 177
Molecular Function: cell adhesion molecule immunoglobulin superfamily cell adhesion molecule receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.