Invitrogen
Manufacturer Code:PA558201
Catalog # PIPA558201
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human AGBL4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP/GTP binding protein-like 4; ATP/GTP-binding protein-like 4; CCP6 ATP/GTP-binding protein-like 4 cytosolic carboxypeptidase 6; Cytosolic carboxypeptidase 6
Gene Aliases: AGBL4; CCP6
UniProt ID: (Human) Q5VU57
Entrez Gene ID: (Human) 84871
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.