Proteintech
Manufacturer Code:169901AP150UL
Catalog # 50-173-1313
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AGBL3 Fusion Protein Ag10592 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGBL3 ATP/GTP binding protein like 3; ATP/GTP binding protein-like 3; ATP/GTP-binding protein-like 3; Cytosolic carboxypeptidase 3
Gene Aliases: 2900053G10Rik; 4930431N21Rik; 6530406M24Rik; AGBL3; CCP3; RGD1561995
UniProt ID: (Human) Q8NEM8, (Mouse) Q8CDP0
Entrez Gene ID: (Human) 340351, (Mouse) 76223, (Rat) 500076
Molecular Function: G-protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.