Novus Biologicals
Manufacturer Code:NB1001323
Catalog # NB1001323
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-NPSAELHRSPSLL corresponding to C-Terminus according to NP_114152.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AGAP-3; ANK repeat and PH domain-containing protein 3 ArfGAP with GTPase domain ankyrin repeat and PH domain 3 centaurin-gamma-3 Cnt-g3 CRAG CRAM-associated GTPase CRMP (collapsin response mediator protein) associated FLJ16146 FLJ34452 gamma 3 MR1-interacting protein MRIP-1; Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 3; centaurin, gamma 3; Centaurin-gamma-3; Cnt-g3; CRAG; CRAM-associated GTPase; CRMP (collapsin response mediator protein) associated; MR1-interacting protein; MRIP-1
Gene Aliases: AGAP-3; AGAP3; CENTG3; cnt-g3; CRAG; MRIP-1
UniProt ID: (Human) Q96P47
Entrez Gene ID: (Human) 116988
Molecular Function: G-protein modulator enzyme modulator nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.