Novus Biologicals
Manufacturer Code:NBP151941
Catalog # NBP151941
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-QASLDSIREAVINSQ corresponding to internal region according to NP_055585.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: AGAP-2; AGAP-2 ArfGAP with GTPase domain ankyrin repeat and PH domain 2 arf-GAP with GTPase ANK repeat and PH domain-containing protein 2 centaurin gamma 1 centaurin-gamma-1 CENTG1 Cnt-g1 FLJ16430 GGAP2 GTP-binding and GTPase activating protein 2 GTP-binding and GTPase-activating protein 2 KIAA0167 Phosphatidylinositol-3-kinase enhancer phosphoinositide 3-kinase enhancer PIKEArf GAP with GTP-binding protein-like ANK repeat and PH domains 2; Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 2; Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2; centaurin, gamma 1; Centaurin-gamma-1; Cnt-g1; GGAP2; GTP-binding and GTPase activating protein 2; GTP-binding and GTPase-activating protein 2; Phosphatidylinositol 3-kinase enhancer; phosphatidylinositol-3-kinase enhancer; phosphoinositide 3-kinase enhancer; PIKE
Gene Aliases: AGAP2; CENTG1; GGAP2; KIAA0167; PIKE
UniProt ID: (Human) Q99490
Entrez Gene ID: (Human) 116986
Molecular Function: G-protein modulator enzyme modulator nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.