Invitrogen
Manufacturer Code:PA597441
Catalog # PIPA597441
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human N(4)-(beta-N-acetylglucosaminyl)-L-asparaginase protein (206-346AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGA; Aspartylglucosaminidase; aspartylglucosylamine deaspartylase; Glycosylasparaginase; Glycosylasparaginase alpha chain; Glycosylasparaginase beta chain; N(4)-(beta-N-acetylglucosaminyl)-L-asparaginase; N4-(N-acetyl-beta-glucosaminyl)-L-asparagine amidase
Gene Aliases: AGA; AGU; ASRG; AW060726; GA
UniProt ID: (Human) P20933, (Mouse) Q64191
Entrez Gene ID: (Human) 175, (Mouse) 11593
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.