Novus Biologicals
Manufacturer Code:NBP188865
Catalog # NBP188865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:DELGETTLDAMIMDGTTMDVGAVGDLRRIKNAIGVARKVLEHTTHTLLVGESATTFAQSMGFINEDLSTSASQALHSDWLARN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGU aspartylglucosaminidase aspartylglucosylamine deaspartylase ASRG EC 3.5.1 EC 3.5.1.26 GA glycosylasparaginase N(4)-(beta-N-acetylglucosaminyl)-L-asparaginase N4-(N-acetyl-beta-glucosaminyl)-L-asparagine amidase; Aspartylglucosaminidase; aspartylglucosylamine deaspartylase; Glycosylasparaginase; Glycosylasparaginase alpha chain; Glycosylasparaginase beta chain; N(4)-(beta-N-acetylglucosaminyl)-L-asparaginase; N4-(N-acetyl-beta-glucosaminyl)-L-asparagine amidase
Gene Aliases: AGA; AGU; ASRG; GA
UniProt ID: (Human) P20933
Entrez Gene ID: (Human) 175
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.