R&D Systems
Manufacturer Code:MAB6307
Catalog # MAB6307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 774855 |
Immunogen | <EM>E. coli</EM>-derived recombinant human AG-3 Ile22-Leu166 Accession # Q8TD06 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AG-3; AG3 AG-3 AGR3 anterior gradient homolog 3 (Xenopus laevis) BCMP11 BCMP11AG3 Breast cancer membrane protein 11 hAG-3member 18 PDIA18; Anterior gradient 3 homolog; anterior gradient homolog 3; Anterior gradient protein 3; anterior gradient protein 3 homolog; Breast cancer membrane protein 11; Protein disulfide isomerase family A, member 18
Gene Aliases: AG-3; AG3; AGR3; BCMP11; hAG-3; HAG3; PDIA18; UNQ642/PRO1272
UniProt ID: (Human) Q8TD06
Entrez Gene ID: (Human) 155465
Molecular Function:
oxidoreductase
reductase
surfactant
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.