Novus Biologicals
Manufacturer Code:NBP215307
Catalog # NBP215307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human AGR3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AG-3; AG-3 anterior gradient homolog 3 (Xenopus laevis) BCMP11AG3 Breast cancer membrane protein 11 hAG-3member 18; Anterior gradient 3 homolog; anterior gradient homolog 3; Anterior gradient protein 3; anterior gradient protein 3 homolog; Breast cancer membrane protein 11; Protein disulfide isomerase family A, member 18
Gene Aliases: AG-3; AG3; AGR3; BCMP11; hAG-3; HAG3; PDIA18; UNQ642/PRO1272
UniProt ID: (Human) Q8TD06
Entrez Gene ID: (Human) 155465
Molecular Function: oxidoreductase reductase surfactant
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.