Invitrogen
Manufacturer Code:PA542924
Catalog # PIPA542924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human AFG3L2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AFG3 ATPase family gene 3-like 2; AFG3 ATPase family gene 3-like 2 (S. cerevisiae) FLJ25993 SCA28; AFG3 ATPase family member 3-like 2; AFG3 like AAA ATPase 2; AFG3(ATPase family gene 3)-like 2; AFG3-like AAA ATPase 2; AFG3-like protein 2; AFG3-like protein 2 (Paraplegin-like protein); ATPase family gene 3, yeast; Paraplegin-like protein
Gene Aliases: 2310036I02Rik; AFG3L2; AW260507; Emv66; par; SCA28; si:ch211-12e1.4; SPAX5
UniProt ID: (Bovine) Q2KJI7, (Human) Q9Y4W6, (Mouse) Q8JZQ2
Entrez Gene ID: (Goat) 102179059, (Dog) 490560, (Horse) 100058377, (Bovine) 515757, (Rabbit) 100350006, (Human) 10939, (Mouse) 69597, (Rat) 307350, (Guinea pig) 100733197, (Zebrafish) 569168
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.