Novus Biologicals
Manufacturer Code:NBP15957220UL
Catalog # NBP15957220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to AFG3L2 (AFG3 ATPase family gene 3-like 2 (yeast) The peptide sequence was selected from the middle region of AFG3L2. Peptide sequence VNFLKNPKQYQDLGAKIPKGAILTGPPGTGKTLLAKATAGEANVPFITVS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AFG3 (ATPase family gene 3 yeast)-like 2 AFG3 ATPase family gene 3-like 2 (S. cerevisiae) AFG3 ATPase family gene 3-like 2 (yeast) AFG3-like protein 2 ATPase family gene 3 yeast EC 3.4.24 EC 3.4.24.- FLJ25993 Paraplegin-like protein SCA28 spinocerebellar ataxia 28; AFG3 ATPase family gene 3-like 2; AFG3 ATPase family member 3-like 2; AFG3 like AAA ATPase 2; AFG3-like AAA ATPase 2; AFG3-like protein 2; ATPase family gene 3, yeast; Paraplegin-like protein
Gene Aliases: AFG3L2; SCA28; SPAX5
UniProt ID: (Human) Q9Y4W6
Entrez Gene ID: (Human) 10939
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.