Proteintech
Manufacturer Code:146311AP150UL
Catalog # 50-172-9600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AFG3L2 Fusion Protein Ag6209 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AFG3 ATPase family gene 3-like 2; AFG3 ATPase family member 3-like 2; AFG3 like AAA ATPase 2; AFG3 like protein 2 AFG3L2 Paraplegin like protein; AFG3(ATPase family gene 3)-like 2; AFG3-like AAA ATPase 2; AFG3-like protein 2; ATPase family gene 3, yeast; Paraplegin-like protein
Gene Aliases: 2310036I02Rik; AFG3L2; AW260507; Emv66; par; SCA28; SPAX5
UniProt ID: (Human) Q9Y4W6, (Mouse) Q8JZQ2
Entrez Gene ID: (Human) 10939, (Mouse) 69597, (Rat) 307350
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.