Proteintech
Manufacturer Code:146621AP150UL
Catalog # 50-172-9625
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AFF4 Fusion Protein Ag6336 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AF4/FMR2 family member 4; AF4/FMR2 family member 4 AF4/FMR2 family member 4 AF5Q31 AFF4 MCEF Protein AF 5q31; AF4/FMR2 family, member 4; ALL1 fused gene from 5q31; ALL1-fused gene from chromosome 5q31 protein; Major CDK9 elongation factor-associated protein; Protein AF-5q31
Gene Aliases: AF5Q31; AFF4; Alf4; CHOPS; HSPC092; Laf4l; MCEF
UniProt ID: (Human) Q9UHB7, (Mouse) Q9ESC8
Entrez Gene ID: (Human) 27125, (Rat) 303132, (Mouse) 93736
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.