Invitrogen
Manufacturer Code:MA531614
Catalog # PIMA531614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2H6D1 |
Immunogen | Purified recombit fragment of human AFAP1L2 (AA: 674-818) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: actin filament associated protein 1-like 2; Actin filament-associated protein 1-like 2; AFAP1-like protein 2; CTB-1144G6.6; XB130 KIAA1914 CTB-1144G6.4
Gene Aliases: AFAP1L2; CTB-1144G6.4; KIAA1914; XB130
UniProt ID: (Human) Q8N4X5
Entrez Gene ID: (Human) 84632
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.