Proteintech
Manufacturer Code:171831AP150UL
Catalog # 50-173-1427
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AFAP1L2 Fusion Protein Ag10966 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actin filament associated protein 1-like 2; Actin filament-associated protein 1-like 2; AFAP1 like protein 2 AFAP1L2 KIAA1914 XB130; AFAP1-like protein 2; CTB-1144G6.6; phosphatidylinositol 3-kinase-associated protein
Gene Aliases: AFAP1L2; AU041783; C86904; CTB-1144G6.4; KIAA1914; mKIAA1914; RGD1310668; XB130
UniProt ID: (Human) Q8N4X5, (Mouse) Q5DTU0
Entrez Gene ID: (Human) 84632, (Rat) 292130, (Mouse) 226250
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.