OriGene
Manufacturer Code:TA810440
Catalog # 50-168-3394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3G4 |
Immunogen | Full length human recombit protein of human AFAP1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 110 kDa actin filament-associated protein; Actin filament-associated protein 1; actin filament-associated protein, 110 kDa; AFAP AFAP-110 AFAP110; AFAP-110
Gene Aliases: AFAP; AFAP-110; AFAP1; AFAP110
UniProt ID: (Human) E9PDT7
Entrez Gene ID: (Human) 60312
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.