Invitrogen
Manufacturer Code:PA544796
Catalog # PIPA544796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human AFAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 110 kDa actin filament-associated protein; Actin filament associated protein 1 AFAP AFAP-110 FLJ56849 AFAP1 AFAP110; Actin filament-associated protein 1; actin filament-associated protein, 110 kDa; AFAP-110
Gene Aliases: 2600003E23Rik; 9630044L16Rik; AFAP; AFAP-110; AFAP1; AFAP110; AI848729; im:7148276; Kiaa3018; mKIAA3018
UniProt ID: (Human) E9PDT7, (Mouse) Q80YS6, (Rat) Q8VH46
Entrez Gene ID: (Bovine) 534032, (Dog) 488786, (Horse) 100056653, (Rabbit) 100339958, (Human) 60312, (Guinea pig) 100735141, (Mouse) 70292, (Rat) 140935, (Zebrafish) 568356
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.