R&D Systems
Manufacturer Code:AF4558
Catalog # AF4558
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human AFAP His450-Thr730 Accession # Q8N556 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 110 kDa actin filament-associated protein; actin filament associated protein 1 AFAP1 AFAP-110 FLJ56849; Actin filament-associated protein 1; actin filament-associated protein, 110 kDa; AFAP-110
Gene Aliases: AFAP; AFAP-110; AFAP1; AFAP110
UniProt ID: (Human) E9PDT7
Entrez Gene ID: (Human) 60312
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.