Invitrogen
Manufacturer Code:PA527797
Catalog # PIPA527797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 281 and 532 of Human AF9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AF9 YEATS3 ALL1-fused gene from chromosome 9 protein YEATS domain-containing protein 3 myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog) translocated to 3 myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein protein AF-9; ALL1-fused gene from chromosome 9 protein; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 3; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 3; Myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein; myeloid/lymphoid or mixed-lineage leukemia; translocated to 3; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 3; Protein AF-9; YEATS domain-containing protein 3
Gene Aliases: AF9; MLLT3; YEATS3
UniProt ID: (Human) P42568
Entrez Gene ID: (Human) 4300
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.