Novus Biologicals
Manufacturer Code:NB1001566F
Catalog # NB1001566F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 525 and the C-terminus (residue 568) of human ALL1 Fused Gene from Chromosome 9 Protein using the numbering given in entry NP_004520.1 (GeneID 4300). |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AF-9 AF9myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog)translocated to 3 ALL1-fused gene from chromosome 9 protein myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog) translocatedto 3 myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog Drosophila)translocated to 3 Myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein protein AF-9 YEATS domain-containing protein 3 YEATS3FLJ2035; ALL1-fused gene from chromosome 9 protein; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 3; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 3; Myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein; myeloid/lymphoid or mixed-lineage leukemia; translocated to 3; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 3; Protein AF-9; YEATS domain-containing protein 3
Gene Aliases: AF9; MLLT3; YEATS3
UniProt ID: (Human) P42568
Entrez Gene ID: (Human) 4300
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.