Proteintech
Manufacturer Code:138601AP150UL
Catalog # 50-172-9048
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AEG-1/MTDH-Specific Fusion Protein Ag4840 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3D3 3D3/LYRIC AEG 1 AEG1 LYRIC LYRIC/3D3 metadherin Metastasis adhesion protein MTDH Protein LYRIC; 3D3/LYRIC; AEG-1; astrocyte elevated gene 1; Astrocyte elevated gene-1 protein; LYRIC; Lysine-rich CEACAM1 co-isolated protein; Metadherin; Metastasis adhesion protein; Protein LYRIC
Gene Aliases: 2610103J23Rik; 3D3; 3D3/Lyric; AEG-1; AEG1; AV353288; D8Bwg1112e; LYRIC; LYRIC/3D3; MTDH
UniProt ID: (Human) Q86UE4, (Mouse) Q80WJ7, (Rat) Q9Z1W6
Entrez Gene ID: (Human) 92140, (Mouse) 67154, (Rat) 170910
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.