Invitrogen
Manufacturer Code:MA524267
Catalog # PIMA524267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 590831 |
Immunogen | Mouse myeloma cell line NS0-derived recombit human ACLP Glu345-Phe1158 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACLP AE-binding protein 1 adipocyte enhancer binding protein 1 adipocyte enhancer-binding protein 1 aortic carboxypeptidase-like protein; Adipocyte enhancer-binding protein 1; AE-binding protein 1; Aortic carboxypeptidase-like protein
Gene Aliases: ACLP; AEBP1
UniProt ID: (Human) Q8IUX7
Entrez Gene ID: (Human) 165
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.