Invitrogen
Manufacturer Code:PA5109366
Catalog # PIPA5109366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 999-1158 of human AEBP1 (NP_001120.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACLP Adipocyte enhancer-binding protein 1 AE-binding protein 1 AEBP1 Aortic carboxypeptidase-like protein BOS_4375 FLJ33612; Adipocyte enhancer-binding protein 1; AE-binding protein 1; Aortic carboxypeptidase-like protein
Gene Aliases: ACLP; AEBP1
UniProt ID: (Human) Q8IUX7, (Rat) A2RUV9, (Mouse) Q640N1
Entrez Gene ID: (Human) 165, (Rat) 305494, (Mouse) 11568
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.