Invitrogen
Manufacturer Code:PA525767
Catalog # PIPA525767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 96-123 amino acids from the N-terminal region of human ADSSL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenylosuccinate synthetase 1, muscle; Adenylosuccinate synthetase isozyme 1; Adenylosuccinate synthetase like 1; Adenylosuccinate synthetase, basic isozyme; Adenylosuccinate synthetase, muscle isozyme; Adenylosuccinate synthetase-like 1; adSS 1; AdSSL1; AMPSase 1; AMPSase 1 IMP--aspartate ligase 1 M-type adenylosuccinate synthetase adSS 1 adenylosuccinate synthetase isozyme 1 adenylosuccinate synthetase basic isozyme adenylosuccinate synthetase muscle isozyme; IMP--aspartate ligase 1; M-type adenylosuccinate synthetase
Gene Aliases: Adss; ADSS1; ADSSL1; AI528595
UniProt ID: (Human) Q8N142, (Mouse) P28650
Entrez Gene ID: (Human) 122622, (Mouse) 11565
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.