Invitrogen
Manufacturer Code:PA555070
Catalog # PIPA555070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ADSS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenylosuccinate synthetase (Ade(-)H-complementing); adenylosuccinate synthetase 2, non muscle; Adenylosuccinate synthetase isozyme 2; Adenylosuccinate synthetase, acidic isozyme; Adenylosuccinate synthetase, liver isozyme; adenylosuccinate synthetase, non muscle; adenylosuccinate synthetase, non-muscle isozyme; adSS 2; AMPSase 2; IMP--aspartate ligase 2; L-type adenylosuccinate synthetase
Gene Aliases: ADEH; ADSS; ADSS 2; ADSS2; AI314886; AS
UniProt ID: (Human) P30520, (Mouse) P46664
Entrez Gene ID: (Human) 159, (Mouse) 11566, (Rat) 289276
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.